missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Mesoporphyrin IX dihydrochloride, 97%

Manufacturer:  Alfa Aesar™ 43320MD

 View more versions of this product

Catalog No. AA43320MD



Specifications

Specifications

Mesoporphyrin IX dihydrochloride
68938-72-7
MFCD00136049
YUIUEEROGVTICN-UHFFFAOYSA-N
3-[18-(2-carboxyethyl)-8,13-diethyl-3,7,12,17-tetramethyl-22,23-dihydroporphyrin-2-yl]propanoic acid;dihydrochloride
111858
Crystalline
250mg
C34H40Cl2N4O4
21h,23h-porphine-2,18-dipropanoic acid, 7,12-diethyl-3,8,13,17-tetramethyl-, dihydrochloride
CCC1=C(C2=CC3=C(C(=C(N3)C=C4C(=C(C(=N4)C=C5C(=C(C(=N5)C=C1N2)C)CCC(=O)O)CCC(=O)O)C)C)CC)C.Cl.Cl
639.618
639.62
97%

Chemical Identifiers

68938-72-7
639.618
YUIUEEROGVTICN-UHFFFAOYSA-N
111858
CCC1=C(C2=CC3=C(C(=C(N3)C=C4C(=C(C(=N4)C=C5C(=C(C(=N5)C=C1N2)C)CCC(=O)O)CCC(=O)O)C)C)CC)C.Cl.Cl
C34H40Cl2N4O4
MFCD00136049
21h,23h-porphine-2,18-dipropanoic acid, 7,12-diethyl-3,8,13,17-tetramethyl-, dihydrochloride
3-[18-(2-carboxyethyl)-8,13-diethyl-3,7,12,17-tetramethyl-22,23-dihydroporphyrin-2-yl]propanoic acid;dihydrochloride
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents