missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
meso-Erythritol, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC117825000
Specifications
(10% in H2O) clear, colorless | |
Authentic | |
1% max. (K.F.) | |
White | |
500g | |
149-32-6 | |
C4H10O4 | |
MFCD00004710 | |
15,3733 | |
(10% in water) Clear colorless | |
C(C(C(CO)O)O)O | |
122.12 | |
CHEBI:17113 | |
Crystalline Powder or Crystals |
meso-Erythritol | |
Plastic bottle | |
329.0°C to 331.0°C | |
117.0°C to 121.0°C | |
99% | |
98.5% min. (ELSD) (HPLC) | |
HOCH2[CH(OH)]2CH2OH | |
01,525 | |
2r,3s-butane-1,2,3,4-tetrol, erythrit, erythrite, erythritol, erythro-tetritol, erythrol, meso-erythritol, mesoerythritol, phycite, phycitol | |
UNXHWFMMPAWVPI-ZXZARUISSA-N | |
(2S,3R)-butane-1,2,3,4-tetrol | |
222285 | |
122.12 | |
99% |
Chemical Identifiers
149-32-6 | |
122.12 | |
UNXHWFMMPAWVPI-ZXZARUISSA-N | |
222285 | |
(2S,3R)-butane-1,2,3,4-tetrol |
C4H10O4 | |
MFCD00004710 | |
2r,3s-butane-1,2,3,4-tetrol, erythrit, erythrite, erythritol, erythro-tetritol, erythrol, meso-erythritol, mesoerythritol, phycite, phycitol | |
CHEBI:17113 | |
C(C(C(CO)O)O)O |
Safety and Handling
EINECSNumber : 205-737-3
RTECSNumber : KF2000000