missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
meso-2,3-Dimercaptosuccinic acid, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 226670050
Specifications
meso-2, 3-Dimercaptosuccinic acid | |
1% max. (1 g, 105°C, 3 hrs) | |
0.2% max. | |
196°C to 198°C | |
98% | |
98% | |
HO2CCH(SH)CH(SH)CO2H | |
15,8995 | |
ACTRVOBWPAIOHC-XIXRPRMCSA-N | |
(2S,3R)-2,3-bis(sulfanyl)butanedioic acid | |
2724354 | |
182.21 | |
98% |
Authentic | |
Glass bottle | |
White | |
5g | |
304-55-2 | |
C4H6O4S2 | |
MFCD00064799 | |
2r,3s-rel-2,3-dimercaptosuccinic acid, chemet, dim-sa, dimercaptosuccinic acid, dmsa, meso-2,3-dimercaptosuccinic acid, meso-dimercaptosuccinic acid, succimer, succimero, succimerum | |
C(C(C(=O)O)S)(C(=O)O)S | |
182.21 | |
CHEBI:63623 | |
Powder |
Chemical Identifiers
304-55-2 | |
182.21 | |
ACTRVOBWPAIOHC-XIXRPRMCSA-N | |
2724354 | |
(2S,3R)-2,3-bis(sulfanyl)butanedioic acid |
C4H6O4S2 | |
MFCD00064799 | |
2r,3s-rel-2,3-dimercaptosuccinic acid, chemet, dim-sa, dimercaptosuccinic acid, dmsa, meso-2,3-dimercaptosuccinic acid, meso-dimercaptosuccinic acid, succimer, succimero, succimerum | |
CHEBI:63623 | |
C(C(C(=O)O)S)(C(=O)O)S |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility or the unborn child.
GHS Signal Word: Warning
EINECSNumber : 206-155-2
RTECSNumber : WM7650000
TSCA : TSCA