missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

meso-2,3-Dimercaptosuccinic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC226671000

 View more versions of this product

Catalog No. AC226671000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

meso-2, 3-Dimercaptosuccinic acid
1% max. (1 g, 105°C, 3 hrs)
0.2% max.
196°C to 198°C
98%
98%
HO2CCH(SH)CH(SH)CO2H
15,8995
ACTRVOBWPAIOHC-XIXRPRMCSA-N
(2S,3R)-2,3-bis(sulfanyl)butanedioic acid
2724354
182.21
98%
Authentic
Glass bottle
White
100g
304-55-2
C4H6O4S2
MFCD00064799
2r,3s-rel-2,3-dimercaptosuccinic acid, chemet, dim-sa, dimercaptosuccinic acid, dmsa, meso-2,3-dimercaptosuccinic acid, meso-dimercaptosuccinic acid, succimer, succimero, succimerum
C(C(C(=O)O)S)(C(=O)O)S
182.21
CHEBI:63623
Powder

Chemical Identifiers

304-55-2
182.21
ACTRVOBWPAIOHC-XIXRPRMCSA-N
2724354
(2S,3R)-2,3-bis(sulfanyl)butanedioic acid
C4H6O4S2
MFCD00064799
2r,3s-rel-2,3-dimercaptosuccinic acid, chemet, dim-sa, dimercaptosuccinic acid, dmsa, meso-2,3-dimercaptosuccinic acid, meso-dimercaptosuccinic acid, succimer, succimero, succimerum
CHEBI:63623
C(C(C(=O)O)S)(C(=O)O)S
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility or the unborn child.

GHS Signal Word: Warning

EINECSNumber : 206-155-2

RTECSNumber : WM7650000

TSCA : TSCA

SDS Documents