Please login to your online account to display your discounted pricing

meso-2,3-Dibromosuccinic acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  147570050

 View more versions of this product

Catalog No. AC147570050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 275.88
Color White to pale yellow to beige
Quantity 5g
Packaging Glass Bottle
Chemical Name or Material meso-2, 3-Dibromosuccinic acid
Assay Percent Range 98%
Linear Formula HO2CCH(Br)CH(Br)CO2H
CAS 608-36-6
Infrared Spectrum Authentic
Molecular Formula C4H4Br2O4
Health Hazard 1 Warning

Specifications

Chemical Name or Material meso-2, 3-Dibromosuccinic acid
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H4Br2O4
Linear Formula HO2CCH(Br)CH(Br)CO2H
Formula Weight 275.88
Accessories

Warning

SDS & Safety Documents