Please login to your online account to display your discounted pricing

Alfa Aesar™ (Mesitylene)tungsten tricarbonyl, 98%

Chemical Identifiers

Chemical Name or Material (Mesitylene)tungsten tricarbonyl
CAS 12129-69-0
Formula Weight 388.08
MDL Number MFCD00015319
UN Number UN3466
Synonym Tricarbonyl(mesitylene)tungsten
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3958903

  SDS
alfa aesar™
3958903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39589MD

  SDS
alfa aesar™
39589MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 388.08
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 12129-69-0
Health Hazard 1 Danger
Assay 98%
MDL Number MFCD00015319
Physical Form Crystalline
Chemical Name or Material (Mesitylene)tungsten tricarbonyl
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
Melting Point 165°C (dec.)
Quantity 1g
UN Number UN3466
Synonym Tricarbonyl(mesitylene)tungsten
Health Hazard 2 GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.