Learn More
Mesitylene, 97%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 161322500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Mesitylene | |
-45.0°C | |
0.8675g/mL | |
44°C | |
96% min. (GC) | |
C9H12 | |
C6H3(CH3)3 | |
250 mL | |
0.8675 | |
mesitylene, sym-trimethylbenzene, benzene, 1,3,5-trimethyl, fleet-x, trimethylbenzol, 3,5-dimethyltoluene, s-trimethylbenzene, hsdb 92, 2,4,6-trimethylbenzene, 1,3,5-trimethyl-benzene | |
AUHZEENZYGFFBQ-UHFFFAOYSA-N | |
1,3,5-trimethylbenzene | |
7947 | |
CHEBI:34833 | |
97% | |
Liquid |
108-67-8 | |
Colorless | |
163.0°C to 166.0°C | |
Authentic | |
Glass bottle | |
1.4980 to 1.5000 | |
MFCD00008538 | |
05, 406 | |
15, 5977 | |
Solubility in water: 2.9g/L (20°C). Other solubilities: miscible with alcohol,benzene and ether | |
CC1=CC(=CC(=C1)C)C | |
120.19 | |
0.834 mm2/s (20°C) | |
120.19 | |
Pure |
Chemical Identifiers
108-67-8 | |
120.19 | |
AUHZEENZYGFFBQ-UHFFFAOYSA-N | |
7947 | |
1,3,5-trimethylbenzene |
C9H12 | |
MFCD00008538 | |
mesitylene, sym-trimethylbenzene, benzene, 1,3,5-trimethyl, fleet-x, trimethylbenzol, 3,5-dimethyltoluene, s-trimethylbenzene, hsdb 92, 2,4,6-trimethylbenzene, 1,3,5-trimethyl-benzene | |
CHEBI:34833 | |
CC1=CC(=CC(=C1)C)C |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.<br
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
GHS Signal Word: Danger
EINECSNumber : 203-604-4
RUO – Research Use Only