Please login to your online account to display your discounted pricing

Alfa Aesar™ Mesitylene-2-sulfonyl hydrazide, 97%

Chemical Identifiers

Chemical Name or Material Mesitylene-2-sulfonyl hydrazide
CAS 16182-15-3
Formula Weight 214.28
MDL Number MFCD00007585
UN Number UN1325
Beilstein 2118380
Synonym 2,4,6-Trimethylbenzenesulfonyl hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5558603

  SDS
alfa aesar™
H5558603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5558606

  SDS
alfa aesar™
H5558606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5558614

  SDS
alfa aesar™
H5558614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 214.28
Melting Point 112° to 114°C (dec.)
Physical Form Powder
Assay 97%
CAS 16182-15-3
Beilstein 2118380
Color White to off-white
Quantity 1g
Chemical Name or Material Mesitylene-2-sulfonyl hydrazide
MDL Number MFCD00007585
UN Number UN1325
Synonym 2,4,6-Trimethylbenzenesulfonyl hydrazide
DOT Information Transport Hazard Class: 4.1; Packaging Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.