missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Mesitylene-2-sulfonyl hydrazide, 97%

Manufacturer:  Alfa Aesar™ H5558614

 View more versions of this product

Catalog No. AAH5558614



Specifications

Specifications

Mesitylene-2-sulfonyl hydrazide
112°C to 114°C (decomposition)
16182-15-3
MFCD00007585
2118380
JQUBKTQDNVZHIY-UHFFFAOYSA-N
2,4,6-trimethylbenzenesulfonohydrazide
85321
Powder
White
25g
C9H14N2O2S
UN1325
2,4,6-trimethylbenzenesulfonyl hydrazide
CC1=CC(=C(C(=C1)C)S(=O)(=O)NN)C
214.283
214.28
97%

Chemical Identifiers

16182-15-3
214.283
JQUBKTQDNVZHIY-UHFFFAOYSA-N
85321
CC1=CC(=C(C(=C1)C)S(=O)(=O)NN)C
C9H14N2O2S
MFCD00007585
2,4,6-trimethylbenzenesulfonyl hydrazide
2,4,6-trimethylbenzenesulfonohydrazide
Safety and Handling

Safety and Handling

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335-H500

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 240-319-4

TSCA : No

Recommended Storage : Ambient temperatures

Documents