missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Mesitylene-2-sulfonyl chloride, 99%

Manufacturer:  Alfa Aesar™ A1177536

 View more versions of this product

Catalog No. AAA1177536


 

Description

Description

Applications
Mesitylene-2-sulfonyl chloride is used as reagent for intramolecular lactamizations and lactonization. It also acts as a reagent for protecting guanidino groups and tryptophan. It is used as coupling reagent in polynucleotide synthesis

Solubility
Soluble in toluene (almost transparency).

Notes
Moisture Sensitive. Store under inert gas. Incompatible with strong bases. Store away from moisture.
Specifications

Specifications

Mesitylene-2-sulfonyl chloride
99%
MFCD00007434
1107601
PVJZBZSCGJAWNG-UHFFFAOYSA-N
2,4,6-trimethylbenzenesulfonyl chloride
150°C (20mmHg)
13046
>110°C (230°F)
Irritating
773-64-8
C9H11ClO2S
UN3261
2,4,6-Trimethylbenzenesulfonyl chloride
CC1=CC(=C(C(=C1)C)S(=O)(=O)Cl)C
53°C to 57°C
218.695
500g
218.7

Chemical Identifiers

773-64-8
218.695
PVJZBZSCGJAWNG-UHFFFAOYSA-N
13046
C9H11ClO2S
MFCD00007434
2,4,6-Trimethylbenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : DB8930000

Recommended Storage : 2°C to 8°C

SDS Documents