missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Mesitylacetic acid, 98+%

$44.41 - $431.80

Chemical Identifiers

CAS 4408-60-0
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00014368
InChI Key CQWMQAKKAHTCSC-UHFFFAOYSA-N
Synonym 2-mesitylacetic acid, mesitylacetic acid, 2,4,6-trimethylphenylacetic acid, 2,4,6-trimethylphenyl acetic acid, 2-2,4,6-trimethylphenyl acetic acid, mesityleneacetic acid, 2,4,6-trimethylbenzeneacetic acid, 2-mesityleneacetic acid, benzeneacetic acid, 2,4,6-trimethyl, 2,4,6-trimethyl-phenyl-acetic acid
PubChem CID 78123
IUPAC Name 2-(2,4,6-trimethylphenyl)acetic acid
SMILES CC1=CC(=C(C(=C1)C)CC(=O)O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0033403 View Documents Alfa Aesar™
L0033403
1g Each for $44.41
 
AAL0033406 View Documents Alfa Aesar™
L0033406
5g Each for $129.54
 
AAL0033414 View Documents Alfa Aesar™
L0033414
25g Each for $431.80
 
Specifications

Chemical Identifiers

4408-60-0
178.231
CQWMQAKKAHTCSC-UHFFFAOYSA-N
78123
CC1=CC(=C(C(=C1)C)CC(=O)O)C
C11H14O2
MFCD00014368
2-mesitylacetic acid, mesitylacetic acid, 2,4,6-trimethylphenylacetic acid, 2,4,6-trimethylphenyl acetic acid, 2-2,4,6-trimethylphenyl acetic acid, mesityleneacetic acid, 2,4,6-trimethylbenzeneacetic acid, 2-mesityleneacetic acid, benzeneacetic acid, 2,4,6-trimethyl, 2,4,6-trimethyl-phenyl-acetic acid
2-(2,4,6-trimethylphenyl)acetic acid

Specifications

167°C to 170°C
4408-60-0
MFCD00014368
2-mesitylacetic acid, mesitylacetic acid, 2,4,6-trimethylphenylacetic acid, 2,4,6-trimethylphenyl acetic acid, 2-2,4,6-trimethylphenyl acetic acid, mesityleneacetic acid, 2,4,6-trimethylbenzeneacetic acid, 2-mesityleneacetic acid, benzeneacetic acid, 2,4,6-trimethyl, 2,4,6-trimethyl-phenyl-acetic acid
CC1=CC(=C(C(=C1)C)CC(=O)O)C
178.231
178.23
Mesitylacetic acid
1g
C11H14O2
2209880
CQWMQAKKAHTCSC-UHFFFAOYSA-N
2-(2,4,6-trimethylphenyl)acetic acid
78123
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning