missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 125600010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Mesityl oxide | |
-53°C | |
129°C | |
Authentic | |
Glass Bottle | |
1.4420 to 1.446 | |
1 L | |
0.854 | |
mesityl oxide, 4-methyl-3-penten-2-one, 3-penten-2-one, 4-methyl, isopropylideneacetone, isobutenyl methyl ketone, methyl isobutenyl ketone, mesityloxid, mesityloxyde, 3-isohexen-2-one, isopropylidene acetone | |
SHOJXDKTYKFBRD-UHFFFAOYSA-N | |
4-methylpent-3-en-2-one | |
8858 | |
98.14 | |
Liquid |
141-79-7 | |
0.8540g/mL | |
28°C | |
90% alpha-Isomer (GC) | |
C6H10O | |
(CH3)2C=CHCOCH3 | |
01,736 | |
15,5978 | |
Solubility in water: 28g/L (20°C). Other solubilities: miscible with most common organic solvents | |
CC(=CC(=O)C)C | |
98.14 | |
0.64 mPa.s (20°C) | |
99% |
Chemical Identifiers
141-79-7 | |
98.14 | |
mesityl oxide, 4-methyl-3-penten-2-one, 3-penten-2-one, 4-methyl, isopropylideneacetone, isobutenyl methyl ketone, methyl isobutenyl ketone, mesityloxid, mesityloxyde, 3-isohexen-2-one, isopropylidene acetone | |
4-methylpent-3-en-2-one |
C6H10O | |
SHOJXDKTYKFBRD-UHFFFAOYSA-N | |
8858 | |
CC(=CC(=O)C)C |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 205-502-5
Recommended Storage : Product may darken during storage
RUO – Research Use Only