missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mesityl oxide, 90+%, remainder 4-methyl-4-penten-2-one, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals A15111AP

 View more versions of this product

Catalog No. AAA15111AP



Description

Description

Mesityl oxide is used as a solvent and in the production of methyl isobutyl ketone by hydrogenation, in the synthesis of 2,3,4,5-tetrahydro-1,3,3-trimethyldipyrrin, a crucial building block in the rational synthesis of chlorins and oxochlorins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mesityl oxide is used as a solvent and in the production of methyl isobutyl ketone by hydrogenation, in the synthesis of 2,3,4,5-tetrahydro-1,3,3-trimethyldipyrrin, a crucial building block in the rational synthesis of chlorins and oxochlorins.

Solubility
Soluble in water(30g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

Mesityl oxide
-59°C
129°C to 130°C
≥90%
remainder 4-methyl-4-penten-2-one
1.444
500 mL
1361550
mesityl oxide, 4-methyl-3-penten-2-one, 3-penten-2-one, 4-methyl, isopropylideneacetone, isobutenyl methyl ketone, methyl isobutenyl ketone, mesityloxid, mesityloxyde, 3-isohexen-2-one, isopropylidene acetone
SHOJXDKTYKFBRD-UHFFFAOYSA-N
4-methylpent-3-en-2-one
8858
≥90%
141-79-7
0.856
30°C (86°F)
honigähnlich
C6H10O
MFCD00008900
UN1229
14,5908
Soluble in water(30g/L).
CC(=CC(=O)C)C
98.145
98.15
Liquid

Chemical Identifiers

141-79-7
98.145
SHOJXDKTYKFBRD-UHFFFAOYSA-N
8858
CC(=CC(=O)C)C
C6H10O
MFCD00008900
mesityl oxide, 4-methyl-3-penten-2-one, 3-penten-2-one, 4-methyl, isopropylideneacetone, isobutenyl methyl ketone, methyl isobutenyl ketone, mesityloxid, mesityloxyde, 3-isohexen-2-one, isopropylidene acetone
4-methylpent-3-en-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H312-H332
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c

H226-H302+H312+H332

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MESITYL OXIDE

EINECSNumber : 205-502-5

RTECSNumber : SB4200000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only