missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mesitaldehyde, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 125570500
Specifications
Mesitaldehyde | |
105°C | |
Glass bottle | |
1 | |
Colorless to Yellow | |
50g | |
487-68-3 | |
C10H12O | |
MFCD00003341 | |
2,4,6-trimethyl-benzaldehyde, 2-formylmesitylene, 2-mesitylenecarboxaldehyde, b-isodural, benzaldehyde, 2,4,6-trimethyl, isodural, mesitaldehyde, mesityl aldehyde, mesitylaldehyde, mesitylenecarboxaldehyde | |
CC1=CC(=C(C(=C1)C)C=O)C | |
148.2 | |
148.2 | |
97% |
1.0000g/mL | |
Authentic | |
1.5520 to 1.5540 | |
237.0°C | |
14.0°C | |
98% | |
96% min. (GC) | |
(CH3)3C6H2CHO | |
07, 325 | |
HIKRJHFHGKZKRI-UHFFFAOYSA-N | |
2,4,6-trimethylbenzaldehyde | |
10254 | |
Liquid |
Chemical Identifiers
487-68-3 | |
148.2 | |
HIKRJHFHGKZKRI-UHFFFAOYSA-N | |
10254 | |
CC1=CC(=C(C(=C1)C)C=O)C |
C10H12O | |
MFCD00003341 | |
2,4,6-trimethyl-benzaldehyde, 2-formylmesitylene, 2-mesitylenecarboxaldehyde, b-isodural, benzaldehyde, 2,4,6-trimethyl, isodural, mesitaldehyde, mesityl aldehyde, mesitylaldehyde, mesitylenecarboxaldehyde | |
2,4,6-trimethylbenzaldehyde |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 207-662-1
RTECSNumber : CU8500000
TSCA : TSCA