Learn More
Mesitaldehyde, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1010409
Description
Mesitaldehyde is used in organic synthesis, and also used as medicine and other fine chemical products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMesitaldehyde is used in organic synthesis, and also used as medicine and other fine chemical products.
Solubility
Soluble in Chloroform
Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Mesitaldehyde | |
14°C | |
237°C | |
C10H12O | |
MFCD00003341 | |
1364114 | |
mesitaldehyde, mesitylaldehyde, benzaldehyde, 2,4,6-trimethyl, 2-mesitylenecarboxaldehyde, mesitylenecarboxaldehyde, 2-formylmesitylene, 2,4,6-trimethyl-benzaldehyde, isodural, mesityl aldehyde, b-isodural | |
HIKRJHFHGKZKRI-UHFFFAOYSA-N | |
2,4,6-trimethylbenzaldehyde | |
10254 | |
98% |
487-68-3 | |
1.005 | |
106°C (222°F) | |
1.554 | |
10 g | |
Air Sensitive | |
Soluble in Chloroform | |
CC1=CC(C)=C(C=O)C(C)=C1 | |
148.21 | |
148.21 |
Chemical Identifiers
487-68-3 | |
148.21 | |
HIKRJHFHGKZKRI-UHFFFAOYSA-N | |
10254 | |
CC1=CC(C)=C(C=O)C(C)=C1 |
C10H12O | |
MFCD00003341 | |
mesitaldehyde, mesitylaldehyde, benzaldehyde, 2,4,6-trimethyl, 2-mesitylenecarboxaldehyde, mesitylenecarboxaldehyde, 2-formylmesitylene, 2,4,6-trimethyl-benzaldehyde, isodural, mesityl aldehyde, b-isodural | |
2,4,6-trimethylbenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-662-1
RTECSNumber : CU8500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only