missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Mesitaldehyde, 98%

Manufacturer:  Alfa Aesar™ A1010404

 View more versions of this product

Catalog No. AAA1010404



Description

Description

Applications
Mesitaldehyde is used in organic synthesis, and also used as medicine and other fine chemical products.

Solubility
Soluble in Chloroform

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Mesitaldehyde
237°C
2g
C10H12O
1364114
HIKRJHFHGKZKRI-UHFFFAOYSA-N
2,4,6-trimethylbenzaldehyde
10254
98%
106°C (222°F)
14°C
487-68-3
MFCD00003341
mesitaldehyde, mesitylaldehyde, benzaldehyde, 2,4,6-trimethyl, 2-mesitylenecarboxaldehyde, mesitylenecarboxaldehyde, 2-formylmesitylene, 2,4,6-trimethyl-benzaldehyde, isodural, mesityl aldehyde, b-isodural
CC1=CC(=C(C(=C1)C)C=O)C
148.205
148.21

Chemical Identifiers

487-68-3
148.205
HIKRJHFHGKZKRI-UHFFFAOYSA-N
10254
CC1=CC(=C(C(=C1)C)C=O)C
C10H12O
MFCD00003341
mesitaldehyde, mesitylaldehyde, benzaldehyde, 2,4,6-trimethyl, 2-mesitylenecarboxaldehyde, mesitylenecarboxaldehyde, 2-formylmesitylene, 2,4,6-trimethyl-benzaldehyde, isodural, mesityl aldehyde, b-isodural
2,4,6-trimethylbenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CU8500000

TSCA : TSCA

SDS Documents