Please login to your online account to display your discounted pricing

Alfa Aesar™ MES lysis buffer with NP-40 (2X)

Chemical Identifiers

Chemical Name or Material MES lysis buffer with NP-40
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ60244EQE

  SDS
alfa aesar™
J60244EQE
125mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Concentration 2X
TSCA TSCA
Quantity 125mL
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Chemical Name or Material MES lysis buffer with NP-40
Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.