Please login to your online account to display your discounted pricing

Alfa Aesar™ MES-buffered saline (5X, pH 7.4), low endotoxin

Chemical Identifiers

Chemical Name or Material MES-buffered saline (5X, pH 7.4)
Name Note low endotoxin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67504AE

 
alfa aesar™
J67504AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67504AK

 
alfa aesar™
J67504AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material MES-buffered saline (5X, pH 7.4)
Color Colorless
Quantity 100mL
TSCA TSCA
Name Note low endotoxin
Physical Form Liquid