missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ MES, 1.0M buffer soln., pH 8.0

$133.08 - $205.11

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.248
MDL Number MFCD00283848
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61556AK View Documents Alfa Aesar™
J61556AK
250mL Each for $133.08
 
AAJ61556AP View Documents Alfa Aesar™
J61556AP
500mL Each for $205.11
 
Description

Description

Applications
MES, is often used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures. It is also been used in various protein characterization assays. This sterile solution suitable for cell culture and molecular biology applications. It is also used as a buffering agent in biology and biochemistry.

Solubility
Soluble in water (0.5 M at 20°C).

Notes
Solutions are stable at 2-8°C for months. Sterilize by filtration through 0.2 mm filters. Autoclaving is not recommended for any sulfonic acid buffer. If buffers must be nuclease-free, treat the water first, then add the buffer after autoclaving. When MES solutions are autoclaved, they turn yellow (although pH does not change measurably). The identity of the yellow breakdown product is unknown.
Specifications

Chemical Identifiers

145224-94-8
213.248
MIIIXQJBDGSIKL-UHFFFAOYSA-N
2-morpholin-4-ylethanesulfonic acid;hydrate
C6H15NO5S
MFCD00283848
16218417
C1COCCN1CCS(=O)(=O)O.O

Specifications

145224-94-8
MFCD00283848
MIIIXQJBDGSIKL-UHFFFAOYSA-N
2-morpholin-4-ylethanesulfonic acid;hydrate
16218417
8.0
250mL
C6H15NO5S
1.0M buffer soln.
C1COCCN1CCS(=O)(=O)O.O
213.248
Liquid
Colorless
MES buffer soln.
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA