missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ MES, 0.5M buffer soln., pH 7.5

$66.62 - $99.47

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.248
MDL Number MFCD00283848
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63706AE View Documents Alfa Aesar™
J63706AE
100mL Each for $66.62
 
AAJ63706AK View Documents Alfa Aesar™
J63706AK
250mL Each for $99.47
 
Specifications

Chemical Identifiers

145224-94-8
213.248
MIIIXQJBDGSIKL-UHFFFAOYSA-N
2-morpholin-4-ylethanesulfonic acid;hydrate
C6H15NO5S
MFCD00283848
16218417
C1COCCN1CCS(=O)(=O)O.O

Specifications

145224-94-8
MFCD00283848
MIIIXQJBDGSIKL-UHFFFAOYSA-N
2-morpholin-4-ylethanesulfonic acid;hydrate
16218417
7.5
100mL
C6H15NO5S
0.5M buffer soln.
C1COCCN1CCS(=O)(=O)O.O
213.248
Liquid
Colorless
MES buffer soln.
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning