Please login to your online account to display your discounted pricing

Alfa Aesar™ MES, 0.2M buffer soln., pH 7.0, low endotoxin


Quantity 100mL

Manufacturer: alfa aesar™  J67058AE

 View more versions of this product

Catalog No. AAJ67058AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material MES
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
MDL Number MFCD00283848
CAS 145224-94-8
Name Note low endotoxin
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
TSCA TSCA
Color Colorless
Concentration 0.2M buffer soln.
Physical Form Liquid
Quantity 100mL
pH 7.0
Health Hazard 1 Warning

Specifications

Chemical Name or Material MES
CAS 145224-94-8
Name Note low endotoxin
MDL Number MFCD00283848
Accessories

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents