Please login to your online account to display your discounted pricing

Alfa Aesar™ MES, 0.2M buffer soln., pH 6.5, low endotoxin

Chemical Identifiers

Chemical Name or Material MES
CAS 145224-94-8
Name Note low endotoxin
MDL Number MFCD00283848
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67372AE

 
alfa aesar™
J67372AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67372AK

 
alfa aesar™
J67372AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD00283848
Chemical Name or Material MES
Physical Form Liquid
Name Note low endotoxin
pH 6.5
Concentration 0.2M buffer soln.
TSCA TSCA
Quantity 100mL
Color Colorless
CAS 145224-94-8