Invitrogen™ MERTK (cMER) Recombinant Human Protein DFS Item
MERTK (cMER) is a tyrosine kinase proto-oncogene.
Manufacturer:  Invitrogen™ LSPV4112
 View more versions of this product
Catalog No. PV4112
Description
Description
MERTK (cMER) is involved in the retinal pigment epithelium (RPE) phagocytosis pathway, which is implicated in human retinal disease.
Specifications
Specifications
NP_006334
Frozen
63.4kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
AXL Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MERTK (cMER), MER
Baculovirus
Human
GST Tag
Documents