MERTK (cMER) [A708S] Recombinant Human Protein, Invitrogen™ DFS Item
MERTK (c-MER) is a member of the TYRO3/AXL/MER (TAM) receptor kinase family. Recombinant human MERTK A708S (578-872) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Manufacturer:  Life Technologies LSPV6326
 View more versions of this product
Catalog No. PV6326
Description
Description
MERTK (c-MER) encodes a transmembrane protein with two fibronectin type-III domains, two Ig-like C2-type (immunoglobulin-like) domains, and one tyrosine kinase domain. Cytokine-dependent activation of TAM signaling diverts a proinflammatory pathway to provide an intrinsic feedback inhibitor of both TLR- and cytokine-driven immune responses. MERTK is essential regulators of mammalian spermatogenesis. Mutations in MERTK gene have been associated with disruption of the retinal pigment epithelium (RPE) phagocytosis pathway and onset of autosomal recessive retinitis pigmentosa (RP).
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications
Specifications
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
60.2kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Kinase Assay
Human
GST Tag
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
100μg
MERTK
aa 578-872
Yes
Documents