Please login to your online account to display your discounted pricing

Alfa Aesar™ Merrifield Resin, 1% crosslinked, 200-400 mesh, 1.0-1.3mmol/g

$149.47 - $439.97

Chemical Identifiers

Chemical Name or Material Merrifield Resin
CAS 55844-94-5
Name Note 1% crosslinked
MDL Number MFCD00146406
Synonym Chloromethyl polystyrene; Chloromethylated styrene-divinylbenzene copolymer
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1702714

  SDS
alfa aesar™
L1702714
25g Each for $149.47

AAL1702722

  SDS
alfa aesar™
L1702722
100g Each for $439.97
Description & Specifications

Specifications

Chemical Name or Material Merrifield Resin
MDL Number MFCD00146406
Synonym Chloromethyl polystyrene; Chloromethylated styrene-divinylbenzene copolymer
Recommended Storage 2° to 8°C
Quantity 25g
Concentration 1.0 to 1.3mmol/g
Name Note 1% crosslinked
CAS 55844-94-5
Safety and Handling

2° to 8°C