Please login to your online account to display your discounted pricing

Alfa Aesar™ Meropenem trihydrate, 98-101%

A broad spectrum antibiotic targeting a wide range of bacteria

Chemical Identifiers

Chemical Name or Material Meropenem trihydrate
CAS 96036-03-2
Formula Weight 383.46
MDL Number MFCD08600005
Merck Index 14,5900
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66446MF

  SDS
alfa aesar™
J66446MF
50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 383.46
Chemical Name or Material Meropenem trihydrate
CAS 96036-03-2
Health Hazard 1 Warning
Assay 98 to 101%
MDL Number MFCD08600005
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 627°C
Color Off-white to pale yellow
Storage Note 1 2° to 8°C
Quantity 50mg
RTECS Number CL5446509
Physical Form Powder
Merck Index 14,5900
Solubility Information Soluble in aqueous solution at approximately 5mg/mL

Applications
Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.

Solubility
Soluble in aqueous solution at approximately 5mg/ml

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C