Please login to your online account to display your discounted pricing

Mercury Standard, 1mL = 0.02mg Hg, 20ppm Hg, Ricca Chemical


Quantity 500mL
Packaging Poly natural

Manufacturer: ricca chemical company  RIRTCLPHG500

 View more versions of this product

 Alert

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Catalog No. RTCLPHG-500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 500mL
Chemical Name or Material Mercury Standard
CAS 7732-18-5
CAS 7439-97-6
CAS Max % 0.0016
CAS Min % 91.0
CAS 7697-37-2
Shelf Life 12 months
CAS Min % 0.0016
CAS Max % 95.0
Packaging Poly natural
CAS Min % 6.84
CAS Max % 7.12
Concentration or Composition (by Analyte or Components) 1mL = 0.02mg Hg, 20ppm Hg

Specifications

Chemical Name or Material Mercury Standard
CAS 7732-18-5
CAS Min % 91.0
CAS Max % 95.0
CAS 7697-37-2
CAS Min % 6.84
CAS Max % 7.12
CAS 7439-97-6
CAS Min % 0.0016
CAS Max % 0.0016
Physical Form Liquid
Accessories

12 months