Please login to your online account to display your discounted pricing
Mercury Spill Kit
Offers complete vapor suppression and mercury containment.
Manufacturer: fisher science education  20MKIT3EMD
 View more versions of this product
Catalog No. S41268
  • / Each
Description & Specifications
Specifications
Product Type Complete Mercury Spill Kit
For Use With (Application) For mercury spill containment
Includes Mercury aspirator, 8.5 oz. Mercon spray, Mercontainer™ waste storage container, 8.5 oz. Merconvap liquid decontaminate, pair of safety goggles, two pairs of nitrile gloves, two disposal bags, two mercury waste labels, storage box