missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mercury Spill Kit

Offers complete vapor suppression and mercury containment.

$74.80 - $334.80

Specifications

For Use With (Application) For mercury spill containment
View More Specs

Includes:
  • Mercury aspirator
  • Mercon spray, 8.5 oz. (250mL)
  • Mercontainer™ waste storage container
  • Merconvap liquid decontaminate, 8.5 oz. (250mL)
  • Pair of safety goggles
  • Two pairs of nitrile gloves
  • Two disposal bags
  • Two mercury waste labels
  • Storage box with plastic handle


Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
S41268 Fisher Science Education
20MKIT3EMD
Complete Mercury Spill Kit Each for $334.80
S412861 Fisher Science Education
20MCNT16EMD
Mercontainer™ Each for $74.80
Case of 6 Each for $448.50
Specifications

Description

  • For small mercury spills from thermometers, manometers and other instruments
  • Mercon™ spray reduces airborne mercury vapor prior to cleanup
  • Merconvap™ treats surfaces after removal of mercury

Description & Specifications

Specifications

For mercury spill containment