Please login to your online account to display your discounted pricing

Alfa Aesar™ Mercury(II) oxide, yellow, 98+%

Chemical Identifiers

Chemical Name or Material Mercury(II) oxide, yellow
CAS 21908-53-2
Molecular Formula HgO
Formula Weight 216.59g/mol
MDL Number MFCD00011045
UN Number UN1641
Merck Index 14,5883
Synonym Mercuric oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1410618

 
alfa aesar™
A14106-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA141060B

 
alfa aesar™
A14106-0B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Odorless
Melting Point 500°C (dec.)
Quantity 50g
Chemical Name or Material Mercury(II) oxide, yellow
CAS 21908-53-2
Assay ≥98%
Molecular Formula HgO
Formula Weight 216.59g/mol
MDL Number MFCD00011045
UN Number UN1641
Merck Index 14,5883
Synonym Mercuric oxide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY OXIDE
RTECS Number OW8750000
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.