Please login to your online account to display your discounted pricing

Alfa Aesar™ Mercury(II) nitrate hydrate, ACS, 98.0% min

$76.99 - $76.99

Chemical Identifiers

Chemical Name or Material Mercury(II) nitrate hydrate
CAS 7783-34-8
Purity Grade ACS Grade
Linear Formula Hg(NO3)2·xH2O (x=1 to 2)
Formula Weight 324.60 (Anhydrous)
MDL Number MFCD00149736
UN Number UN1625
Merck Index 14,5880
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1449718

  SDS
alfa aesar™
1449718*
50g Each for $76.99

AA1449730

  SDS
alfa aesar™
1449730
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 324.60 (Anhydrous)
Health Hazard 3 GHS P Statement
P221-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Take any precaution to avoid mixing with combustibles.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Mercury(II) nitrate hydrate
TSCA TSCA
Purity Grade ACS Grade
Physical Form Crystalline
Assay ≤98.0%
Sensitivity Hygroscopic
Linear Formula Hg(NO3)2·xH2O (x=1 to 2)
MDL Number MFCD00149736
CAS 7783-34-8
UN Number UN1625
Merck Index 14,5880
Color White
Melting Point 79°C
Quantity 50g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H330-H272-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May intensify fire
oxidizer.
May cause damage to organs through prolonged or repeated exposure.
Density 4.39g/mL
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURIC NITRATE
Safety and Handling

Danger

GHS P Statement
P221-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Take any precaution to avoid mixing with combustibles.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330-H272-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May intensify fire
oxidizer.
May cause damage to organs through prolonged or repeated exposure.