missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mercury ICP Standard, 1mL = 10mg Hg (10,000ppm Hg), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPHG10KN500

 View more versions of this product

Catalog No. PHG10KN-500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.


Specifications

Specifications

Mercury ICP Standard
82.0,15.37,0.82
Hg
[Hg]
200.592
CHEBI:16170
Poly natural
1 mL = 10 mg Hg
500mL
7439-97-6,7697-37-2,7732-18-5
85.0,15.99,0.85
QSHDDOUJBYECFT-UHFFFAOYSA-N
mercury
23931
Liquid
10,000ppm Hg
Colorless

Chemical Identifiers

7439-97-6
200.592
23931
mercury
Hg
QSHDDOUJBYECFT-UHFFFAOYSA-N
CHEBI:16170
[Hg]
Documents