missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Mercury bis(trifluoromethylsulfonyl)imide

Manufacturer:  Alfa Aesar™ 4715603

 View more versions of this product

Catalog No. AA4715603



Specifications

Specifications

Mercury bis(trifluoromethylsulfonyl)imide
104715-41-5
Hygroscopic
[(CF3SO2)2N]2Hg
UN2923
RLGIDIMUDWEWMA-UHFFFAOYSA-N
bis(trifluoromethylsulfonyl)azanide;mercury(2+)
131865223
Solid
1g
White
C4F12HgN2O8S4
MFCD23380172
mercury di bis trifluoromethylsulfonylimide
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Hg+2]
760.863
760.86

Chemical Identifiers

104715-41-5
760.863
RLGIDIMUDWEWMA-UHFFFAOYSA-N
131865223
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Hg+2]
C4F12HgN2O8S4
MFCD23380172
mercury di bis trifluoromethylsulfonylimide
bis(trifluoromethylsulfonyl)azanide;mercury(2+)
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H314-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

SDS Documents