Please login to your online account to display your discounted pricing

Alfa Aesar™ Mercury bis(trifluoromethylsulfonyl)imide

Chemical Identifiers

Chemical Name or Material Mercury bis(trifluoromethylsulfonyl)imide
CAS 104715-41-5
Linear Formula [(CF3SO2)2N]2Hg
Formula Weight 760.86
MDL Number MFCD23380172
UN Number UN2923
Synonym Mercury bis(trifluoromethanesulfonyl)imide; Mercury bis(trifluoromethanesulfonimide)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4715603

  SDS
alfa aesar™
4715603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4715606

  SDS
alfa aesar™
4715606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA47156MD

  SDS
alfa aesar™
47156MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 760.86
Physical Form Solid
CAS 104715-41-5
MDL Number MFCD23380172
Quantity 1g
Synonym Mercury bis(trifluoromethanesulfonyl)imide; Mercury bis(trifluoromethanesulfonimide)
Chemical Name or Material Mercury bis(trifluoromethylsulfonyl)imide
UN Number UN2923
Health Hazard 1 Danger
Linear Formula [(CF3SO2)2N]2Hg
Health Hazard 2 GHS H Statement
H301-H311-H331-H314-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Hygroscopic
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Color Off-white
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H314-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.