Please login to your online account to display your discounted pricing

Mercury AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Hg), LabChem™

$41.54 - $88.51

Chemical Identifiers

Chemical Name or Material Mercury AA Standard
CAS 7732-18-5
CAS Max % 89.88
CAS 7697-37-2
CAS Max % 9.94
CAS 22852-67-1
CAS Max % 0.18
Name Note 1000ppm (1mL = 1mg Hg)
Purity Grade Certified Grade
Linear Formula Hg(NO3)2·H2O
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC167501

 
lab chem
LC167501
Poly Bottle 500mL Each for $88.51

LC167507

 
lab chem
LC167507
Poly Bottle 125mL Each for $41.54
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 342.62
Chemical Name or Material Mercury AA Standard
UN Number UN3264
CAS Max % 89.88
Identification Passes Test
Concentration 1000ppm ±10ppm (1mL = 1mg Hg)
Color Clear, colorless
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
May cause damage to organs (nervous system, blood, kidneys, intestinal tract) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
Viscosity 0.99cSt
CAS Max % 9.94
Quantity 500mL
Health Hazard 1 Danger
CAS 7697-37-2
CAS Max % 0.18
Solubility Soluble in water
Linear Formula Hg(NO3)2·H2O
Recommended Storage Room Temperature
Density 1.05g/mL
CAS 22852-67-1
CAS 7732-18-5
Name Note 1000ppm (1mL = 1mg Hg)
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear eye protection, protective gloves, protective clothing.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Physical Form Liquid
Decomposition Information Nitrogen oxides; Mercury
Specific Gravity 1.05g/mL
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear eye protection, protective gloves, protective clothing.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.
May cause damage to organs (nervous system, blood, kidneys, intestinal tract) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.

Room Temperature