missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mercury AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Hg), LabChem™

Manufacturer:  LabChem LC167507

 View more versions of this product

Catalog No. LC167507



Specifications

Specifications

Mercury AA Standard
22852-67-1,7697-37-2,7732-18-5
Hg(NO3)2·H2O
UN3264
Liquid
Certified
Soluble in water
0.99cSt
Colorless
1000ppm (1mL = 1mg Hg)
89.88,9.94,0.18
1000ppm ±10ppm (1mL = 1mg Hg)
342.62
Passes Test
Poly Bottle
Traceable to NIST
1.05g/mL
125mL

Chemical Identifiers

22852-67-1
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause damage to organs (nervous system, blood, kidneys, intestinal tract) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear eye protection, protective gloves, protective clothing.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

Documents
Promotions

Promotions