Please login to your online account to display your discounted pricing

Mercury, 1000μg/mL (0.10% w/v), BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

$115.99 - $138.68

Chemical Identifiers

Chemical Name or Material Mercury, 1000μg/mL (0.10% w/v)
Name Note Plasma Standard
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula Hg
Formula Weight 200.59
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003317

 
avantor performance
576804
Poly Bottle 100mL Each for $138.68

02003318

 
avantor performance
645904
Narrow Mouth Poly Bottle 150mL Each for $115.99
Description & Specifications

Specifications

Formula Weight 200.59
Chemical Name or Material Mercury, 1000μg/mL (0.10% w/v)
Molecular Formula Hg
Name Note Plasma Standard
Packaging Poly Bottle
Quantity 100mL
Specific Gravity 1L = 1.00kg
Purity Grade BAKER INSTRA-ANALYZED™ Reagent