Please login to your online account to display your discounted pricing

Mercuric Sulfate TS, Denigs' Reagent, Ricca Chemical

$106.45 - $106.45

Chemical Identifiers

Chemical Name or Material Mercuric Sulfate
Name Note Denigs' Reagent, Test Solution
CAS 7732-18-5
CAS Min % 64.0
CAS Max % 67.0
CAS 7664-93-9
CAS Min % 29.61
CAS Max % 30.82
CAS 21908-53-2
CAS Min % 4.03
View More Specs

 Alert

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

4779-4

 
ricca chemical company
RI47794
Poly natural 120mL Each for $106.45

4779-16

 
ricca chemical company
RI477916
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 120mL
CAS Min % 64.0
CAS Max % 67.0
Chemical Name or Material Mercuric Sulfate
CAS Max % 4.19
Name Note Denigs' Reagent, Test Solution
CAS 7732-18-5
CAS 7664-93-9
Packaging Poly natural
CAS Min % 29.61
CAS Max % 30.82
CAS 21908-53-2
CAS Min % 4.03