missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mercuric Sulfate Solution, 80g/L Red Mercuric Oxide in 6 Normal Sulfuric Acid, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,7664-93-9,21908-53-2
CAS Min % 70.0,21.59,6.21
CAS Max % 73.0,22.47,6.46
Physical Form Liquid
Chemical Name or Material Mercuric Sulfate Solution
View More Specs

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.



Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
4780-16 Ricca Chemical Company
RI478016
Poly natural 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
4780-32 Ricca Chemical Company
RI478032
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7664-93-9,21908-53-2
73.0,22.47,6.46
Mercuric Sulfate Solution
70.0,21.59,6.21
Liquid

Specifications

Liquid
500mL
70.0,6.21,21.59
6.46,22.47,73.0
Mercuric Sulfate Solution
Clear colorless
80g/L Red Mercuric Oxide in 6Normal Sulfuric Acid
21908-53-2,7664-93-9,7732-18-5
Poly natural