missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mercuric-Potassium Iodide TS, Mayer's Reagent, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,7681-11-0,7487-94-7
CAS Min % 92.0,4.61,1.25
CAS Max % 96.0,4.8,1.3
Physical Form Liquid
Chemical Name or Material Mercuric-Potassium Iodide, Mayer's Reagent, Test Solution
View More Specs

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.



Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
4765-32 Ricca Chemical Company
RI476532
Amber Poly 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
4765-16 Ricca Chemical Company
RI476516
Amber Poly 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7681-11-0,7487-94-7
96.0,4.8,1.3
Mercuric-Potassium Iodide, Mayer's Reagent, Test Solution
92.0,4.61,1.25
Liquid

Specifications

Liquid
1L
7487-94-7,7732-18-5,7681-11-0
Amber Poly
Clear colorless
92.0,4.61,1.25
1.3,96.0,4.8
Mercuric-Potassium Iodide, Mayer's Reagent, Test Solution