Please login to your online account to display your discounted pricing

Mercuric Nitrate, Monohydrate, ACS Grade, ≥ 98.0 , LabChem™

$223.43 - $223.43

Chemical Identifiers

Chemical Name or Material Mercuric Nitrate
CAS 7783-34-8
CAS Max % 100
Name Note Monohydrate
Purity Grade ACS Grade
Linear Formula Hg(NO3)2·H2O
Formula Weight 342.62
UN Number UN1625
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC166459

 
lab chem
LC166459
100g Each for $223.43
Description & Specifications

Specifications

Assay ≥98.0
Formula Weight 342.62
Chemical Name or Material Mercuric Nitrate
Identification Passes Test
Name Note Monohydrate
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Assay Percent Notes as Hg(NO3)2·H2O
Solubility Soluble in water
Specific Gravity 4.3g/cm3
Storage Information Room Temperature
Linear Formula Hg(NO3)2·H2O
CAS 7783-34-8
UN Number UN1625
Decomposition Information Mercury; Nitrogen oxides
Color White
Melting Point 70°C
Quantity 100g
Health Hazard 1 Danger
CAS Max % 100
Health Hazard 2 GHS H Statement
Fatal if swallowed, in contact with skin or if inhaled.
May cause damage to organs (kidney, central nervous system) through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement
Do not breathe dust.
Do not get in eyes, on skin, or on clothing.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin: Wash with plenty of soap and water.
Immediately call a poison center/doctor.
Remove immediately all contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Density 4.3g/cm3
Safety and Handling

Danger

GHS P Statement
Do not breathe dust.
Do not get in eyes, on skin, or on clothing.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin: Wash with plenty of soap and water.
Immediately call a poison center/doctor.
Remove immediately all contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Fatal if swallowed, in contact with skin or if inhaled.
May cause damage to organs (kidney, central nervous system) through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.