Please login to your online account to display your discounted pricing

Mercuric Nitrate, For Vanadium, Certified, 0.35g/L, LabChem™

$60.71 - $60.71

Chemical Identifiers

Chemical Name or Material Mercuric Nitrate
CAS 7732-18-5
CAS Max % 99.91
CAS 7697-37-2
CAS Max % 0.05
CAS 22852-67-1
CAS Max % 0.04
Name Note For Vanadium
Purity Grade Certified Grade
Linear Formula Hg(NO3)2·H2O
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC166801

 
lab chem
LC166801
500mL Each for $60.71
Description & Specifications

Specifications

Formula Weight 342.62
Purity Grade Certified Grade
CAS 22852-67-1
Solubility Soluble in water
Quantity 500mL
Concentration 0.35g/L
Name Note For Vanadium
CAS 7732-18-5
Storage Information Room Temperature
CAS Max % 0.04
Health Hazard 2 GHS H Statement
May cause damage to organs (nervous system, blood, kidneys, intestinal tract) through prolonged or repeated exposure.
Identification Passes Test
Suitability Passes Test
CAS Max % 99.91
Packaging Amber Glass
Chemical Name or Material Mercuric Nitrate
Color Clear, colorless
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Get medical advice and attention if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
CAS 7697-37-2
Decomposition Information Nitrogen oxides; Mercury
CAS Max % 0.05
Health Hazard 1 Warning
Physical Form Liquid
Linear Formula Hg(NO3)2·H2O
Safety and Handling

Warning

GHS P Statement
Do not breathe mist, vapors, spray.
Get medical advice and attention if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May cause damage to organs (nervous system, blood, kidneys, intestinal tract) through prolonged or repeated exposure.