Please login to your online account to display your discounted pricing

Mercuric Nitrate, For Chloride, Certified, 0.0250N ±0.0005N (0.013M), LabChem™

$57.34 - $240.69

Chemical Identifiers

Chemical Name or Material Mercuric Nitrate
CAS 7732-18-5
CAS Max % 99.35
CAS 22852-67-1
CAS Max % 0.45
CAS 7697-37-2
CAS Max % 0.2
Name Note For Chloride
Purity Grade Certified Grade
Linear Formula Hg(NO3)2·H2O
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC166571

 
lab chem
LC166571
500mL Each for $57.34

LC166572

 
lab chem
LC166572
1L Each for $93.77

LC166574

 
lab chem
LC166574
4L Each for $240.69
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 342.62
Chemical Name or Material Mercuric Nitrate
Traceability to NIST Traceable to NIST
CAS Max % 0.2
Decomposition Information Nitrogen oxides; Mercury
Color Clear, colorless
Quantity 500mL
Name Note For Chloride
Physical Form Liquid
Concentration 0.0250N ±0.0005N (0.013M)
Linear Formula Hg(NO3)2·H2O
Density 1g/mL
CAS 7732-18-5
Health Hazard 1 Warning
CAS Max % 99.35
Health Hazard 2 GHS H Statement
Harmful in contact with skin.
May cause damage to organs (nervous system, blood, kidneys) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
Identification Passes Test
CAS 22852-67-1
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Amber Glass
CAS Max % 0.45
Solubility Soluble in water
CAS 7697-37-2
Specific Gravity 1g/mL
Storage Information Room Temperature
Safety and Handling

Warning

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful in contact with skin.
May cause damage to organs (nervous system, blood, kidneys) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.