Please login to your online account to display your discounted pricing

Mercuric Nitrate, For Chloride, Certified, 0.0250N ±0.0005N (0.013M), LabChem™


Quantity 1L

Manufacturer: lab chem  LC166572

 View more versions of this product

Catalog No. LC166572

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Formula Weight 342.62
Linear Formula Hg(NO3)2·H2O
Density 1g/mL
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Amber Glass
Physical Form Liquid
Solubility Soluble in water
Identification Passes Test
CAS 7697-37-2
CAS Max % 0.2
Health Hazard 1 Warning
CAS Max % 99.35
Specific Gravity 1g/mL
CAS 22852-67-1
CAS 7732-18-5
CAS Max % 0.45
Concentration 0.0250N ±0.0005N (0.013M)
Color Clear, colorless
Storage Information Room Temperature
Quantity 1L
Traceability to NIST Traceable to NIST
Health Hazard 2 GHS H Statement
Harmful in contact with skin.
May cause damage to organs (nervous system, blood, kidneys) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
Name Note For Chloride
Chemical Name or Material Mercuric Nitrate
Decomposition Information Nitrogen oxides; Mercury

Specifications

Chemical Name or Material Mercuric Nitrate
CAS 7732-18-5
CAS Max % 99.35
CAS 22852-67-1
CAS Max % 0.45
CAS 7697-37-2
CAS Max % 0.2
Name Note For Chloride
Purity Grade Certified Grade
Linear Formula Hg(NO3)2·H2O
Formula Weight 342.62
Accessories

Warning

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful in contact with skin.
May cause damage to organs (nervous system, blood, kidneys) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.