Please login to your online account to display your discounted pricing

Mercuric Nitrate, Certified, 0.100N ±0.001N (0.05M), LabChem™

$117.60 - $264.86

Chemical Identifiers

Chemical Name or Material Mercuric Nitrate
CAS 7732-18-5
CAS Max % 97.7
CAS 22852-67-1
CAS Max % 1.8
CAS 7697-37-2
CAS Max % 0.5
Purity Grade Certified Grade
Linear Formula Hg(NO3)2·H2O
Formula Weight 342.62
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC166582

 
lab chem
LC166582
1L Each for $117.60

LC166584

 
lab chem
LC166584
4L Each for $264.86
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 342.62
Chemical Name or Material Mercuric Nitrate
CAS Max % 0.5
Identification Passes Test
Packaging Amber Glass
Physical Form Liquid
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Linear Formula Hg(NO3)2·H2O
CAS 7732-18-5
Decomposition Information Nitrogen oxides; Mercury
Color Clear, colorless
Quantity 1L
Health Hazard 1 Warning
CAS Max % 97.7
Health Hazard 2 GHS H Statement
Harmful if swallowed or if inhaled.
May cause damage to organs (nervous system, blood, kidneys) through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
CAS 22852-67-1
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wash exposed skin after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 1.8
Concentration 0.100N ±0.001N (0.05M)
Traceability to NIST Traceable to NIST
Density 1g/mL
CAS 7697-37-2
Safety and Handling

Warning

GHS P Statement
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wash exposed skin after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed or if inhaled.
May cause damage to organs (nervous system, blood, kidneys) through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.