Please login to your online account to display your discounted pricing

Mercuric Nitrate, (1mL = 5mg Cl-), for Chloride, Certified, 0.141N ±0.002N (0.071M), LabChem™

$63.77 - $297.14

Chemical Identifiers

Chemical Name or Material Mercuric Nitrate
CAS 7732-18-5
CAS Max % 96.46
CAS 22852-67-1
CAS Max % 2.54
CAS 7697-37-2
CAS Max % 1
Name Note (1mL = 5mg Cl-), for Chloride
Purity Grade Certified Grade
Linear Formula Hg(NO3)2·H2O
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC166601

 
lab chem
LC166601
500mL Each for $63.77

LC166602

 
lab chem
LC166602
1L Each for $100.06

LC166604

 
lab chem
LC166604
4L Each for $297.14
Description & Specifications

Specifications

Formula Weight 342.62
Purity Grade Certified Grade
Health Hazard 1 Warning
CAS Max % 2.54
Traceability to NIST Traceable to NIST
Density 1g/mL
Physical Form Liquid
Solubility Soluble in water
Packaging Amber Glass
Decomposition Information Nitrogen oxides; Mercury
CAS 7697-37-2
Chemical Name or Material Mercuric Nitrate
Specific Gravity 1g/mL
Name Note (1mL = 5mg Cl-), for Chloride
CAS Max % 96.46
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Storage Information Room Temperature
CAS 7732-18-5
CAS Max % 1
Linear Formula Hg(NO3)2·H2O
Color Clear, colorless
Concentration 0.141N ±0.002N (0.071M)
Identification Passes Test
Health Hazard 2 GHS H Statement
Harmful if swallowed or if inhaled.
Causes skin irritation and serious eye irritation.
May cause damage to organs (nervous system, blood, kidneys) through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
CAS 22852-67-1
Quantity 500mL
Safety and Handling

Warning

GHS P Statement
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed or if inhaled.
Causes skin irritation and serious eye irritation.
May cause damage to organs (nervous system, blood, kidneys) through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.