Please login to your online account to display your discounted pricing

Mercuric Nitrate, (1mL = 1mg NaCl), Certified, 0.0171N ±0.0002N (0.0086M), LabChem™

$74.74 - $166.49

Chemical Identifiers

Chemical Name or Material Mercuric Nitrate
CAS 7732-18-5
CAS Max % 99.59
CAS 22852-67-1
CAS Max % 0.31
CAS 7697-37-2
CAS Max % 0.1
Name Note (1mL = 1mg NaCl)
Purity Grade Certified Grade
Linear Formula Hg(NO3)2·H2O
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC166552

 
lab chem
LC166552
1L Each for $74.74

LC166554

 
lab chem
LC166554
4L Each for $166.49
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 342.62
Identification Passes Test
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.
Specific Gravity 1g/mL
CAS 22852-67-1
Health Hazard 1 Warning
Decomposition Information Nitrogen oxides; Mercury
Color Clear, colorless
Solubility Soluble in water
Packaging Amber Glass
Concentration 0.0171N ±0.0002N (0.0086M)
CAS Max % 0.1
Storage Information Room Temperature
Linear Formula Hg(NO3)2·H2O
Quantity 1L
CAS 7697-37-2
Name Note (1mL = 1mg NaCl)
CAS Max % 0.31
CAS Max % 99.59
Physical Form Liquid
Density 1g/mL
CAS 7732-18-5
Health Hazard 2 GHS H Statement
Harmful in contact with skin.
May cause damage to organs (nervous system, liver, kidneys) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
Chemical Name or Material Mercuric Nitrate
Safety and Handling

Warning

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful in contact with skin.
May cause damage to organs (nervous system, liver, kidneys) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.