missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mercuric Nitrate, (1mL = 0.5mg Cl-), for Chloride, Certified, 0.0141N ±0.0002N (0.0071M), LabChem™

$37.37 - $163.86

Chemical Identifiers

CAS 7732-18-5,22852-67-1,7697-37-2
CAS Max % 99.65,0.25,0.1
Linear Formula Hg(NO3)2·H2O
Formula Weight 342.62
Chemical Name or Material Mercuric Nitrate, (1mL = 0.5mg Cl-), for Chloride
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC166501 Lab Chem
LC166501
500mL Each for $37.37
LC166502 Lab Chem
LC166502
1L Each for $58.66
LC166504 Lab Chem
LC166504
4L Each for $163.86
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,22852-67-1,7697-37-2
Hg(NO3)2·H2O
Mercuric Nitrate, (1mL = 0.5mg Cl-), for Chloride
99.65,0.25,0.1
342.62
Certified Grade

Specifications

7732-18-5,22852-67-1,7697-37-2
Liquid
Hg(NO3)2·H2O
Passes Test
Soluble in water
Traceable to NIST
Clear, colorless
Mercuric Nitrate, (1mL = 0.5mg Cl-), for Chloride
99.65,0.25,0.1
0.0141N ±0.0002N (0.0071M)
342.62
Amber Glass
Room Temperature
Nitrogen oxides; Mercury
500mL
Certified Grade

Safety and Handling

GHS H Statement
Harmful in contact with skin.
May cause damage to organs (nervous system) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Do not breathe mist, vapors, spray.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.

Warning