Please login to your online account to display your discounted pricing

Mercuric Nitrate, 0.0100 Normal, Ricca Chemical

$198.76 - $198.76

Chemical Identifiers

Chemical Name or Material Mercuric Nitrate
Name Note 0.0100 N (N/100)
CAS 7697-37-2
CAS 7732-18-5
CAS 7783-34-8
Physical Form Liquid
Assay Percent Range 0.01000 ± 0.00001 N at 20°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R470400020F

 
ricca chemical company
RIR470400020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R47040004C

 
ricca chemical company
R47040004C
Amber glass 4L Each for $198.76
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 20L
Solvent or Matrix Aqueous Solution
Chemical Name or Material Mercuric Nitrate
Name Note 0.0100 N (N/100)
CAS 7697-37-2
Odor Odorless
Packaging Cubitainer
Assay Percent Range 0.01000 ± 0.00001 N at 20°C
Solubility Solubility in Water: Infinite
CAS 7732-18-5
CAS 7783-34-8