Please login to your online account to display your discounted pricing

Mercuric Chloride, Saturated, Ricca Chemical™


Quantity 500mL
Packaging Glass amber

Manufacturer: ricca chemical company  RIR4670000500C

 View more versions of this product

 Alert

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Catalog No. R4670000500

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500mL
Physical Form Liquid
Color Clear colorless
CAS Max % 6.0
CAS Min % 92.0
CAS Max % 96.0
Chemical Name or Material Mercuric Chloride
Shelf Life 24 months
Name Note Saturated
Packaging Glass amber
CAS 7732-18-5
CAS 7487-94-7
CAS Min % 5.77

Specifications

Chemical Name or Material Mercuric Chloride
Name Note Saturated
CAS 7732-18-5
CAS Min % 92.0
CAS Max % 96.0
CAS 7487-94-7
CAS Min % 5.77
CAS Max % 6.0
Physical Form Liquid
Accessories

24 months